Tisk…

Anti-Endostatin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5799-100EA 6910 CZK
BIOV5799-100
Anti-Endostatin Rabbit Polyclonal Antibody
Protilátky
Endostatin has been identified as a C-terminal fragment of Collagen type 18, a recently identified member of a family of collagen-like proteins referred to as multiplexin family Endostatin specifically inhibits proliferation of endothelial cells althoμgh it does not affect the proliferation of EOMA cells. Endostatin also potently inhibits angiogenesis and tumor growth. Endostatin has an important role in endothelial cell adhesion and cytoskeletal organization. Endostatin can be found in vessel walls (elastic fibers) and basement membranes. Recombinant Endosatin expressed in yeast causes G1 arrest of endothelial cells, and endostatin treatment results in apoptosis of HUVE and HMVE cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Endostatin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR