Tisk…

Anti-HSP70 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5859-100EA 6910 CZK
BIOV5859-100
Anti-HSP70 Rabbit Polyclonal Antibody
Protilátky
HSP70 is a class of highly conserved stress proteins. HSP70 is comprised of multiple members, all of which bind ATP in vitro, but are localized within different intracellular compartments. These include: a) the constitutive Hsc70 (or cognate) present within the cytosol/nucleus; b) the highly stress-inducible HSP70 present within the cytosol/ nucleus/nucleolus; c) the constitutive glucose regulated 78 kDa (or BiP) protein present within the lumen of the endoplasmic reticulum; and d) the glucose regulated 75 kDa protein present within the mitochondria. HSP70 is typically not expressed in the cell under normal growth conditions, but is expressed at high levels in the cell experiencing stress. Therefore, detection of HSP70 using an antibody specific for the inducible form is quite useful in ascertaining whether a stress response has occurred in the cell.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR