Tisk…

Anti-Cox-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3361-100EA 7780 CZK
BIOV3361-100
Anti-Cox-1 Rabbit Polyclonal Antibody
COX proteins are membrane-associated heme proteins that have cyclooxygenase and peroxidase activities. These enzymes are targets of NSAID (Nonsteroidal anti-inflammatory drμgs) such as aspirin. Prostaglandins (PGs) formed by the enzymatic activity of COX-1 are primarily involved in the regulation of homeostatic functions throμghout the body, whereas PGs formed by COX-2 primarily mediate pain, fever, and inflammation. COX-1 is constitutively expressed, with particularly high expression in gastrointestinal tissues. COX-2 is induced by cytokines and mitogens and is likely to play a role in inflammatory diseases such as rheumatoid arthritis.

Recommended Dilutions: Western Blot: 1-4 µg/ml; immunoprecipitation, and Immunohistochemistry(5µg/ml

Type: Primary
Antigen: Cox-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR