Tisk…

Anti-BLCAM Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3363-100EA 8120 CZK
BIOV3363-100
Anti-BLCAM Rabbit Polyclonal Antibody
Cell adhesion molecules (CAMs) are a family of closely related cell surface glycoproteins involved in cell-cell interactions during growth and are thoμght to play an important role in embryogenesis and development. BLCAM α and BLCAM β are produced by alternative splicing of the CD22 gene and form a heterodimer that mediates B cell-B cell interactions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 5-10 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: BLCAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR