Tisk…

Anti-Cathepsin G Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3370-200EA 8040 CZK
BIOV3370-200
Anti-Cathepsin G Rabbit Polyclonal Antibody
Protilátky
The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O. The aspartyl protease class is composed of cathepsins D and E. Cathepsin G is in the serine protease class. Most cathepsins are lysosomal and each is involved in cellular metabolism, participating in various events such as peptide biosynthesis and protein degradation. Cathepsin G expression is restricted to bone marrow and early myeloid cells.

Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: Cathepsin G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR