Tisk…

Anti-STOX2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3382-100EA 7780 CZK
BIOV3382-100
Anti-STOX2 Rabbit Polyclonal Antibody
The Storkhead box protein 2 (STOX2) is a winged-helix domain containing transcription factor. STOX2 has been sμggested to be part of a molecular profile unique to stem cells and may play a role in the differentiation of stem cells.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: STOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR