Tisk…

Anti-Hsp90 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3389-100EA 7900 CZK
BIOV3389-100
Anti-Hsp90 Rabbit Polyclonal Antibody
Protilátky
Heat shock proteins (HSPs) are ubiquitously expressed in all organisms. A major function of HSP90 and other HSPs is to act as molecular chaperones. HSP90 forms a complex with glucocorticoid receptor (GR), rendering the non ligand-bound receptor transcriptionally inactive. HSP 90 binds the GR as a heterocomplex composed of either HSP56 or cyclophilin-40, forming an aporeceptor complex. HSP90 also exists as a dimer with other proteins such as p60/sti1 and p23, forming an aporeceptor complex with estrogen and androgen receptors.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 10-20 µg/ml; Immunohistochemistry: frozen sections, 10-20 µg/ml

Type: Primary
Antigen: Hsp90
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR