Tisk…

Anti-Hsp105 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3390-100EA 7900 CZK
BIOV3390-100
Anti-Hsp105 Rabbit Polyclonal Antibody
Protilátky
Heat shock proteins (HSPs) are ubiquitously expressed in all organisms. The HSPs fall into five general families: HSP105, HSP90, HSP70, the low molecular weight HSPs, and the immunophilins. The HSP105 (also called HSP110) family is composed of HSP105, Apg-1 and Apg-2. The HSP90 family consists of HSP90α, HSP90β and GRP94. The HSP70 family is comprised of HSP70, HSC70, GRP78 and GRP75. The low molecular weight HSPs includes HSP10, HSP25, HSP27 and HSP32. HSP56 and FKBP12 are immunophilins and bind to immunosuppressive drμgs.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: Hsp105
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR