Tisk…

Anti-Etk/BMX Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3398-100EA 7470 CZK
BIOV3398-100
Anti-Etk/BMX Rabbit Polyclonal Antibody
Protilátky
Etk is a member of the Bruton’s tyrosine kinase family. Etk is expressed in a variety of hematopoietic, epithelial and endothelial cells. It participates in multiple signal transduction pathways. Phosphorylation of tyrosine 566 by Src kinase is required for activation of Etk in vivo. In endothelial and epithelial cells, Etk is regulated by FAK throμgh phosphorylation at tyrosine 40.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Etk/BMX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR