Tisk…
Anti-6xHis Mouse Monoclonal Antibody [clone: G020]
Dodavatel: BioVision
BIOV3646-100EA 9390 CZK
BIOV3646-100
Anti-6xHis Mouse Monoclonal Antibody [clone: G020]
It is expected that the His-Tag monoclonal antibody will react with proteins tagged with His from all species.
Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunocytochemistry: 10-20 µg/ml
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: G020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: All Species