Tisk…

Anti-Phospho-MARCKS Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3650-100EA 9730 CZK
BIOV3650-100
Anti-Phospho-MARCKS Rabbit Polyclonal Antibody
Protilátky
MARCKS (Myristoylated alanine-rich protein kinase C substrate) is a major PKC substrate that is distributed in various cell types. MARCKS has been implicated in cell motility, cell adhesion, phagocytosis, membrane traffic and mitogenesis. PKC phosphorylates Ser152, 156 and 163 of MARCKS, which regulates MARCKS's calcium/calmodulin binding activity and filamentous (F)-actin cross-linking activity. In addition, phosphorylation by PKC results in translocation of MARCKS from membrane to cytoplasm.

Recommended Dilutions: Western Blot: 1:200-1000

Type: Primary
Antigen: Phospho-MARCKS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR