Tisk…

Anti-ApoLipoprotein B Goat Polyclonal Antibody

Dodavatel: BioVision
BIOV3661-100EA 8120 CZK
BIOV3661-100
Anti-ApoLipoprotein B Goat Polyclonal Antibody
Apolipoprotein B consists of a single polypeptide chain with a molecular weight of 549 kDa and is mostly synthesised in the liver. It is a major apolipoprotein of very low density, intermediate density and low density lipoproteins (LDL) as well as being a major component of lipoprotein (a;Apolipoprotein B is a ligand for the LDL receptor and elevated levels are associated with premature atherosclerosis. Normal plasma apolipoprotein B levels are around 800 mg/L.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Immunoprecipitation: 5-10 µg/ml; Immunohistochemistry: 5-10 µg/ml; ELISA: 0.5-1 µg/ml

Type: Primary
Antigen: ApoLipoprotein B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR