Tisk…

Anti-MST-1/Krs-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3667-100EA 6910 CZK
BIOV3667-100
Anti-MST-1/Krs-2 Rabbit Polyclonal Antibody
Protilátky
MST-1 (Mammalian STE20-like kinase 1) is a serine/threonine kinase that has been implicated to play a role in chromatin condensation. Its C-terminal contains nuclear export signals which are released upon caspase mediated cleavage. The N-terminus portion of MST-1 then translocates to the nucleus and will promote chromatin condensation. The full length of MST-1 is localized to the cytoplasm.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MST-1/Krs-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR