Tisk…

Anti-Bcl-Rambo Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3671-100EA 7780 CZK
BIOV3671-100
Anti-Bcl-Rambo Rabbit Polyclonal Antibody
Bcl-rambo shares the common structural characteristics with other members of the anti-apoptotic Bcl-2 family members, but differs from them at its C-terminus, where a 250 amino acid sequence proceeds the membrance anchor region. It also differs from other pro-apoptotic Bcl-2 family members in that it membrane anchor C-terminus region is responsible for its apoptotic activity, not its Bcl-2 homology motifs.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Bcl-Rambo
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR