Tisk…

Anti-Bin1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3672-100EA 6910 CZK
BIOV3672-100
Anti-Bin1 Rabbit Polyclonal Antibody
Protilátky
BIN-1 is a tumor suppressor protein that was identified in a genetic screen for proteins that interact with the MCY oncoprotein. BIN-1 inhibits the oncogenic activity of MYC, possibly throμgh activating or facilitating apoptosis. BIN is a short-lived protein with a half-life of about 2 hours.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Bin1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR