Tisk…

Anti-Phospho-BRCA1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3674-100EA 9170 CZK
BIOV3674-100
Anti-Phospho-BRCA1 Rabbit Polyclonal Antibody
Protilátky
BRCA1 is a breast cancer susceptibility gene that was localized to chromosome 17q. Mutations within this gene are believed to account for approximately 45% of families with high incidence of breast cancer and at least 80% of families with increased incidence of both early-onset breast cancer and ovarian cancer. The BRCA1 gene is expressed in numerous tissues, including breast and ovary, and encodes a predicted protein of 1863 amino acids. Like many other genes involved in familial cancer, BRCA1 appears to encode a tumor suppressor, a protein that acts as a negative regulator of tumor growth.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: Phospho-BRCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR