Tisk…

Anti-TAB1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3682-100EA 8430 CZK
BIOV3682-100
Anti-TAB1 Rabbit Polyclonal Antibody
TAB1 (TAK1 binding protein 1) is an adaptor protein that can activate TAK1 in vivo throμgh phosphorylation of TAK1, a mitogen-activated protein kinase kinase kinase. Once activated TAK1 phosphorylates MAPK kinase MKK4 and MKK3/6, which activate p38 MAPK and JNK, respectively.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: TAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR