Tisk…

Anti-MEF2A Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3683-100EA 8430 CZK
BIOV3683-100
Anti-MEF2A Rabbit Polyclonal Antibody
MEF2A is a member of MEF2 (myocyte enhancer factor 2) family of transcription factors. P38 phosphorylates MEF2A at Thr312 and Thr319 within the transcription activation domain and these phosphorylation events promote MEF2A –MEF2D heterodimer dependent gene expression.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: MEF2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR