Tisk…

Anti-PAK3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3686-100EA 6910 CZK
BIOV3686-100
Anti-PAK3 Rabbit Polyclonal Antibody
Protilátky
The p21-activated kinase (PAK) family of serine/threonine kinases is engaged in multiple cellular processes, including cytoskeletal recognization, MAPK signaling, apoptotic signaling, control of phagocytes, NADPH oxidase and growth factor-induced neurite outgrowth. Research indicates that phosphorylation of serine 139 of PAK3 which is located in the kinase inhibitory domain, affects the kinase activity.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PAK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR