Tisk…

Anti-Placental Lactogen Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5869-100EA 7780 CZK
BIOV5869-100
Anti-Placental Lactogen Rabbit Polyclonal Antibody
Human Placental Lactogen (HPL), also called human chorionic somatomammotropin, is a polypeptide placental hormone. Its structure and function is similar to that of growth hormone. It modifies the metabolic state of the mother during pregnancy to facilitate the energy supply of the fetus. Recombinant human Placental Lactogen is a 22.3 kDa protein.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Placental Lactogen
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR