Tisk…

Anti-TFF1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5893-100EA 6910 CZK
BIOV5893-100
Anti-TFF1 Rabbit Polyclonal Antibody
Protilátky
The Trefoil Factor peptides (TFF1, TFF2 and TFF3) are expressed in the gastrointestinal tract, and appear to play an important role in intestinal mucosal defense and repair.

  • Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: TFF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR