Tisk…

Anti-C5a Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5895-100EA 6910 CZK
BIOV5895-100
Anti-C5a Rabbit Polyclonal Antibody
Protilátky
Mouse complement C5a is a 77-amino acid peptide generated during complement activation from the alpha-chain of complement C5. Mouse C5a shares 60% and 82% amino acid sequence identity to human and rat C5a, respectively. C5a binds to a signaling G-protein coupled receptor (GPCR) (C5aR/CD88), inducing neutrophil chemotaxis and endothelial cell activation. It also triggers an oxidative burst in macrophages and neutrophils, and induces release of histamine in basophils and mast cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: C5a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR