Tisk…

Anti-Heregulin- beta 2/Neuregulin-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6645-100EA 7470 CZK
BIOV6645-100
Anti-Heregulin- beta 2/Neuregulin-1 Rabbit Polyclonal Antibody
Protilátky
Neuregulin/Heregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3 and NRG4;To date, there are over 14 soluble and transmembrane proteins derived from the NRG1 gene. Proteolytic processing of the extracellular domain of the transmembrane NRG1 isoforms release soluble growth factors. HRG-β2 is a signaling protein for ErbB2/ErbB4 receptor on the cardiac muscle cells. The polypeptide is comprised of an immunoglobulin and epidermal growth factor-like (EGF-like) domains.

Recommended Dilutions: Western Blot: ~1:200

Type: Primary
Antigen: Heregulin- beta 2/Neuregulin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR