Tisk…

Anti-SCF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6646-100EA 7760 CZK
BIOV6646-100
Anti-SCF Rabbit Polyclonal Antibody
Protilátky
Stem cell factor / KIT ligand (SCF) is a cytokine which binds CD117(c-Kit;SCF is also known as "steel factor" or "c-kit ligand". SCF exists in two forms; cell surface bound SCF and soluble (or free) SCF. Soluble SCF is produced by the cleavage of surface bound SCF by metalloproteases. SCF is a growth factor important for the survival, proliferation, and differentiation of hematopoietic stem cells and other hematopoietic progenitor cells. One of its roles is to change the BFU-E (burst-forming unit-erythroid) cells, which are the earliest erythrocyte precursors in the erythrocytic series, into the CFU-E (colony-forming unit-erythroid).

Recommended Dilutions: Western Blot: ~1:200

Type: Primary
Antigen: SCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR