Tisk…

Anti-Chicken Growth Hormone Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6648-100EA 7330 CZK
BIOV6648-100
Anti-Chicken Growth Hormone Rabbit Polyclonal Antibody
Protilátky
GH is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. It is a 191-amino acid, single chain polypeptide hormone which is synthesized, stored, and secreted by the somatotroph cells within the lateral wings of the anterior pituitary gland. The gene, along with four other related genes, is located at the growth hormone locus on chromosome 17 where they are interspersed in the same transcriptional orientation; an arrangement which is thought to have evolved by a series of gene duplications. The five genes share a remarkably high degree of sequence identity. Alternative splicing generates additional isoforms of each of the five growth hormones, leading to further diversity and potential for specialization. This particular family member is expressed in the pituitary but not in placental tissue as is the case for the other four genes in the growth hormone locus.

Recommended Dilutions: Western Blot: ~1:200

Type: Primary
Antigen: Chicken Growth Hormone
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR