Tisk…

Anti-Neuregulin/Heregulin-1 beta Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6650-100EA 7470 CZK
BIOV6650-100
Anti-Neuregulin/Heregulin-1 beta Rabbit Polyclonal Antibody
Protilátky
Neuregulin/Heregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3 and NRG4;To date, there are over 14 soluble and transmembrane proteins derived from the NRG1 gene. Proteolytic processing of the extracellular domain of the transmembrane NRG1 isoforms release soluble growth factors. HRG1-β1 contains an Ig domain and an EGF-like domain that is necessary for direct binding to receptor tyrosine kinases erb3 and erb4. This binding induces erb3 and erb4 heterodimerization with erb2, stimulating intrinsic kinase activity, which leads to tyrosine phosphorylation. Although HRG1-β1 biological effects is still unclear, it has been found to promote motility and invasiveness of breast cancer cells which may also involve up-regulation of expression and function of the autocrine motility-promoting factor (AMF;Recombinant human Heregulin-β1 (HRG1-β1) is a 7.5 kDa polypeptide consisting of only the EGF domain of Heregulin-β1 (65 amino acid residues).

Recommended Dilutions: Western Blot: ~1:200

Type: Primary
Antigen: Neuregulin/Heregulin-1 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR