Tisk…

Anti-Adenosine Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6652-100EA 9870 CZK
BIOV6652-100
Anti-Adenosine Rabbit Polyclonal Antibody
Adenosine (ADO) is a purine nucleoside comprising a molecule of adenine attached to a ribose sugar molecule (ribofuranose) moiety via a β-N9-glycosidic bond. Adenosine plays an important role in biochemical processes, such as energy transfer — as adenosine triphosphate (ATP) and adenosine diphosphate (ADP) — as well as in signal transduction as cyclic adenosine monophosphate, cAMP. It is also an inhibitory neurotransmitter, believed to play a role in promoting sleep and suppressing arousal.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Adenosine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mammals Samples
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR