Tisk…

Anti-Guanosine Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6654-100EA 8320 CZK
BIOV6654-100
Anti-Guanosine Rabbit Polyclonal Antibody
Protilátky
Guanosine is a purine nucleoside comprising of a guanine attached to a ribose (ribofuranose) ring via a β-N9-glycosidic bond. Guanosine can be phosphorylated to become guanosine monophosphate (GMP), cyclic guanosine monophosphate (cGMP), guanosine diphosphate (GDP), and guanosine triphosphate (GTP;These forms play important roles in various biochemical processes such as synthesis of nucleic acids and proteins, photosynthesis, muscle contraction, and intracellular signal transduction (cGMP;When guanine is attached by its N9 nitrogen to the C1 carbon of a deoxyribose ring it is known as deoxyguanosine.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Guanosine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mammals Samples
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR