Tisk…

Anti-SPHKAP Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6656-100EA 7760 CZK
BIOV6656-100
Anti-SPHKAP Rabbit Polyclonal Antibody
Protilátky
Anchoring protein that binds preferentially to the type I regulatory subunit of c-AMP-dependent protein kinase (PKA type I) and targets it to distinct subcellular compartments. May act as a converging factor linking cAMP and sphingosine signaling pathways. Plays a regulatory role in the modulation of SPHK1.

Recommended Dilutions: Western Blot: 1-4 µg/ml.

Type: Primary
Antigen: SPHKAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR