Tisk…

Anti-GHBP Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6660-100EA 7760 CZK
BIOV6660-100
Anti-GHBP Rabbit Polyclonal Antibody
Protilátky
GHBP is a transmembrane receptor for growth hormone. Binding of growth hormone to the receptor leads to receptor dimerization and the activation of an intra- and intercellular signal transduction pathway leading to growth. A common alternate allele of this gene, called GHRd3, lacks exon three and has been well-characterized. Mutations in this gene have been associated with Laron syndrome, also known as the growth hormone insensitivity syndrome (GHIS), a disorder characterized by short stature. Human Recombinant GHBP expressed from E. coli is a single, non-glycosylated, polypeptide chain containing 237 amino acids and having a molecular mass of 30.3 kDa.

Recommended Dilutions: Western Blot: 1-4 µg/ml.

Type: Primary
Antigen: GHBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR