Tisk…

Anti-TDRD12-BD2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6664-100EA 8880 CZK
BIOV6664-100
Anti-TDRD12-BD2 Rabbit Polyclonal Antibody
Protilátky
Tudor domains are small protein structural motifs of about ~50 amino acids related to the “royal family” of methyl readers, which also includes chromo, MBT, PWWP, and Agenet-like domains. Tudor domains occur either alone, in tandem, or with other domains and are found in many proteins that are involved in RNA metabolism, germ cell development, transposon silencing, DNA damage response, histone modification, and chromatin remodeling. The tudor domains recognize symmetric methylated arginine or methylated lysine residues. Tudor domain proteins act as an oncogene and play a very important role in HCC and colon cancer. TDRD is also involved in RISC complex and interacts with AEG-1 oncogene. The tudor domain can bind to methylated arginine protein and promote tumor angiogenesis in human hepatocellular carcinoma, etc.

Recommended Dilutions: Western Blot: 1-4 µg/ml.

Type: Primary
Antigen: TDRD12-BD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR