Tisk…

Anti-BRD3-BD1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6665-100EA 8320 CZK
BIOV6665-100
Anti-BRD3-BD1 Rabbit Polyclonal Antibody
Protilátky
The acetylation of histone lysine residues plays a crucial role in the epigenetic regulation of gene transcription. A bromodomain is a protein domain that recognizes acetylated lysine residues such as those on the N-terminal tails of histones. This recognition is often a prerequisite for protein-histone association and chromatin remodeling. These domains function in the linking of protein complexes to acetylated nucleosomes, thereby controlling chromatin structure and gene expression. Thus, bromodomains serve as “readers” of histone acetylation marks regulating the transcription of target promoters. BRD3 binds hyper-acetylated chromatin and plays a role in the regulation of transcription, probably by chromatin remodeling and interaction with transcription factors. It regulates transcription by promoting the binding of the transcription factor GATA1 to its targets and transcription of the CCND1 gene. A chromosomal aberration involving BRD3 was found in a rare, aggressive, and lethal carcinoma arising in midline organs of young people.

Recommended Dilutions: Western Blot: 1-4 µg/ml.

Type: Primary
Antigen: BRD3-BD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR