Tisk…

Anti-Cathepsin L Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6669-100EA 7470 CZK
BIOV6669-100
Anti-Cathepsin L Rabbit Polyclonal Antibody
Protilátky
Cathepsin L, a lysosomal endopeptidase expressed in most eukaryotic cells, is a member of the papain-like family of cysteine proteinases.1-3 Cathepsin L plays a major role in antigen processing, tumor invasion and metastasis, bone resorption, and turnover of intracellular and secreted proteins involved in growth regulation.4-6 Although commonly recognized as a lysosomal protease, cathepsin L is also secreted. This broad-spectrum protease is potent in degrading several extracellular proteins (laminins, fibronectin, collagens I and IV, elastin, and other structural proteins of basement membranes) as well as serum proteins and cytoplasmic and nuclear proteins.

Recommended Dilutions: Western Blot: 1-4 µg/ml.

Type: Primary
Antigen: Cathepsin L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR