Tisk…

Anti-Cyr61/CCN1 Rat Monoclonal Antibody [clone: 466305]

Dodavatel: BioVision
BIOV6696-50EA 10570 CZK
BIOV6696-50
Anti-Cyr61/CCN1 Rat Monoclonal Antibody [clone: 466305]
Protilátky
Cyr61 is a member of angiogenic and vasculogenic regulators designated CCN proteins, which includes CTGF, Cyr61 homolog, Fisp2. Cyr61 functions as an angiogenic inducer, stimulating cell adhesion and migration and promotes DNA synthesis of human vascular endothelial cells. Elevated expression of Cyr61 is observed during vessel growth, wound healing and chondrocyte differentiation.

Recommended Dilutions: Western Blot: 2 µg/ml, IHC: 8-25 µg/ml.

Type: Primary
Antigen: Cyr61/CCN1
Clonality: Monoclonal
Clone: 466305
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2B
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR