Tisk…

Anti-Phospho-p38 MAPK Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3438-100EA 9730 CZK
BIOV3438-100
Anti-Phospho-p38 MAPK Rabbit Polyclonal Antibody
Protilátky
p38 MAP kinase is the mammalian homologue of the yeast HOG kinase and participates in a
cascade controlling cellular responses to cytokines and stress. Like the SAPK/JNK pathway,
p38 MAP kinase is activated by a variety of cellular stresses including inflammatory cytokines,
UV light and growth factors, etc. Activated p38 MAP kinase has been shown to phosphorylate
and activate MAPKAP kinase-2 and to phosphorylate the transcription factors ATF-2 and Max.

Recommended Dilutions: Western Blot: 0.5-4 ?g/ml; Immunoprecipitation: 10-20 mg/ml; Immunohistochemistry: 10-20 ?g/ml

Type: Primary
Antigen: Phospho-p38 MAPK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR