Tisk…

Anti-TLR3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3445R-100EA 7780 CZK
BIOV3445R-100
Anti-TLR3 Rabbit Polyclonal Antibody
To date, at least ten members of the Toll family have been identified. TLRs recognize molecular patterns associated with microbial pathogens and induce antimicrobial immune response. TLR3 recognizes double-stranded (ds) RNA, induces the activation of NF-kB, throμgh MyD88-dependent and -independent pathways, and the production of type I interferons (IFNs;Similar to several other members of the TLR family, TLR3 has been reported to be expressed at a very low level on the surface of human fibroblast cell lines.

Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: TLR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR