Tisk…

Anti-Paxillin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3448-100EA 7470 CZK
BIOV3448-100
Anti-Paxillin Rabbit Polyclonal Antibody
Protilátky
Paxillin is one of the key components of integrin signaling. Tyrosine phosphorylation of paxillin is required for integrin-mediated cytoskeletal reorganization. Paxillin is phosphorylated by another focal adhesion component, focal adhesion kinase (FAK), at tyrosine 118. Phospho-Tyr118 may provide a docking site for recruitment of other signaling molecules to focal adhesions.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: Paxillin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR