Tisk…

Anti-PDK1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3449-100EA 8120 CZK
BIOV3449-100
Anti-PDK1 Rabbit Polyclonal Antibody
PDK1 (Phosphoinositide-dependent protein kinase 1) plays an important role in cell signaling pathways and is involved in regulation of a wide variety of processes, including cell proliferation, differentiation and apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 15-25 µg/ml

Type: Primary
Antigen: PDK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR