Tisk…

Anti-PKC Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3450-100EA 7470 CZK
BIOV3450-100
Anti-PKC Rabbit Polyclonal Antibody
Protilátky
PKC (Protein kinase C) is a calcium-dependent and phospholipid-dependent enzyme that is activated in vivo by the lipid diacylglycerol. PKC has been shown to be involved in signal transduction and various aspects of neoplastic transformation. PKC contains an amino terminal regulatory domain and a carboxy terminal catalytic domain joined by a hinge region. Cleavage of protein kinase C in vitro results in the generation of two functional fragments corresponding to these two domains.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PKC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR