Tisk…

Anti-Superoxide Dismutase Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3458-100EA 8120 CZK
BIOV3458-100
Anti-Superoxide Dismutase Rabbit Polyclonal Antibody
SOD (Superoxide Dismutase) is a well characterized cytosolic scavenger of oxygen free radicals that requires copper and zinc binding to potentiate its enzymatic activity. Enzymatically, SOD-1 facilitates the dismutation of oxygen radicals to hydrogen peroxide, and it also catalyzes prooxidant reactions, which include the peroxidase activity and hydroxyl radical generating activity. Defects in the gene encoding SOD-1 have been implicated in the progression of neurological diseases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 20 µg/ml; Immunohistochemistry: 20 µg/ml, frozen & paraffin

Type: Primary
Antigen: Superoxide Dismutase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR