Tisk…
HPLC/UHPLC columns, Avantor® ACE® C18-Amide, 2 µm
Dodavatel: Avantor®
ACE®
ACE-1012-7501EA 18210 CZK
ACE-1012-7501 ACE-1012-10005 ACE-1012-1001S ACE-1012-75005 ACE-1012-1201 ACE-1012-1501S ACE-1012-10005S ACE-1012-1501 ACE-1012-0501 ACE-1012-0301 ACE-1012-1001 ACE-1012-75005S ACE-1012-1201S ACE-1012-0301S ACE-1012-7501S ACE-1012-15005 ACE-1012-05005 ACE-1012-05005S ACE-1012-15005S ACE-1012-12005S ACE-1012-12005 ACE-1012-0501S ACE-1012-03005S ACE-1012-03005
HPLC/UHPLC columns, Avantor® ACE® C18-Amide, 2 µm
Kolony chromatografické
Avantor® ACE® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2,0 to 10 µm) and complementary phases
  • Low bleed for UV and LC/MS compatibility
  • C18 hydrophobic plus Amide interactions, compatible with 100% aqueous conditions
  • Method validation kits contain three different batches of the same phase - available on request
The special design of the ACE® C18-Amide ligand incorporates an extended alkyl chain spacer between the silica surface and amide polar group, which gives extra phase stability.