Tisk…

Rat IgG2b Isotype Control [clone: LTF-2]

Dodavatel: Tonbo Biosciences
40-4031-U500EA 3460 CZK
40-4031-U500
Rat IgG2b Isotype Control [clone: LTF-2]
Protilátky
The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.

Functional Assay

Type: Control
Antigen: IgG2b
Clonality: Monoclonal
Clone: LTF-2
Conjugation: In Vivo Ready™
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR