Tisk…
Mini rotator, digital
Dodavatel: Thermo Fisher Scientific
BARN88880026EA 0 CZK
BARN88880026
Mini rotator, digital
Třepačky Třepačky rotační
This versatile digitally controlled compact rotator provides smooth, quiet horizontal mixing for different tubes/vessels. It is used in bacteria and fungi culture, gel staining and de-staining processes and in hybridisation assays.
  • PID control for consistent and smooth rocking motion
  • Ideal for placing in incubators (non-CO₂), chambers, and refrigerators (up to 60 °C)
  • Displays both current and set RPM simultaneously
Speed acuracy (at 100 min⁻¹): ±5 min⁻¹
Informace pro objednávání: Units are supplied without platforms. Universal platforms and other accessories must be ordered separately.