Tisk…

Anti-RBC Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
ROCK101-4139EA 12430 CZK
ROCK101-4139
Anti-RBC Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR