Tisk…

Anti-RBC Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
ROCK203-4139EA 9580 CZK
ROCK203-4139
Anti-RBC Rabbit Polyclonal Antibody
Protilátky
contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: RBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR