Tisk…

Anti-HDAC11 Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
ROCK600-401-J17EA 10660 CZK
ROCK600-401-J17
Anti-HDAC11 Rabbit Polyclonal Antibody
Protilátky
Histone deacetylases, such as HDAC11, control DNA expression by modifying the core histone octamers that package DNA into dense chromatin structures and repress gene expression. Anti-HDAC11 antibodies are ideal for researchers interested in Breast Cancer, Cancer, Epigenetics, and Histone Deacetylases research.

Type: Primary
Antigen: HDAC11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR