Tisk…

Anti-HDAC2 Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
ROCK600-401-J19EA 10660 CZK
ROCK600-401-J19
Anti-HDAC2 Rabbit Polyclonal Antibody
Protilátky
Histone deacetylase 2 (HDAC2), or transcriptional regulator homolog RPD3 L1, is highly homologous to the yeast transcription factor RPD3 (reduced potassium dependency 3) gene. As in yeast, human HDAC2 is likely to be involved in regulating chromatin structure during transcription. It has been implicated to associate with YY1, a mammalian zinc-finger transcription factor, which negatively regulates transcription by tethering RPD3 to DNA as a cofactor. This process is highly conserved from yeast to human. Anti-HDAC2 antibodies are ideal for researchers interested in Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases, and Stem Cell Markers research.

Type: Primary
Antigen: HDAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Primate
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR