Tisk…
Precision Balances, Cubis® Essentials
Dodavatel: Sartorius
SARBMSE8201SCEED15EA 91200 CZK
SARBMSE8201SCEED15 SARBMSE1203SCEED15 SARBMSE12201SCED15 SARBMSE5201SCEED15 SARBMSE4202SCEED15 SARBMSE5201S-ED15 SARBMSE6202SCEED15 SARBMSE623SCE-ED15
Precision Balances, Cubis® Essentials
High end weighing with monolithic weighing technology for users without complex operations who want to perform precise weighing.
  • MSE display - high contrast, black and white, backlit display
  • Easy to understand menu guidance wth short text prompts
  • Software support for fast levelling
  • Rectangular weighing pans
  • Three optional ports are available as accesory ( Bluetooth®, PS/2, RS232C)
Applications include unit conversion, isoCAL automatic calibration | adjustment function, density determination (buoyancy method only), calculations, averaging, net | total formulation, weighing in percent, counting, totalising.