Tisk…
Test sieves, 200×25 mm
Dodavatel: Endecotts
ENDE200BIW.425HEA 5210 CZK
ENDE200BIW.425H ENDE200BIW.106H ENDE200BIW.100H ENDE200BIW.045H ENDE200BIW.038H ENDE200BIW.125H ENDE200BIW.212H ENDE200BIW.300H ENDE200BIW.500H ENDE200BIW.180H ENDE200BIW.090H ENDE200BIW.250H ENDE200BIW.075H ENDE200BIW.053H ENDE200BIW.063H ENDE200BIW.150H
Test sieves, 200×25 mm
Woven wire mesh test sieves with brass frame and stainless steel mesh are ideal for all types of laboratory sampling and particle size analysis.
For more size options please contact your local Avantor office.
Certifikace: ISO3310-1:2000, BS410-1:2000; DIN-ISO3310-1; NF ISO3310-1:2000
Informace o dodávce: Each sieve is supplied together with a certificate of examination and carries the BS 410 kite mark.