Tisk…
Certification and calibration of test sieves
Dodavatel: VWR, part of Avantor
VWR®
510-1617EA 2960 CZK
510-1617 510-1620 510-1619 510-1618 510-1616 510-1621
Certification and calibration of test sieves
Services
Each test sieve is delivered with a free certificate of compliance with the order 2.1 in accordance with DIN EN 10204. It certifies that the metal wire cloth for test sieve and the test sieve itself are manufactured in compliance with the quality assurance instructions according to ISO 9001.
If required, inspection certificates 3.1 in accordance with DIN EN 10204 for test sieves are also available. They document the distribution of aperture widths in graphic and tubular form.
Tests can be carried out at two confidence levels as a certifying or calibrating measurement. For calibration, three times as many meshes are measured, achieving a particularly high level of statistic reliability. The measurements are finally evaluated separately in the warp and weft directions and the results of the measurements are saved for subsequent check measurements.